Thermo Scientific Chemicals

Methyl 2-aminopyridine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: L19869.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2-aminopyridine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: L19869.06
5 g, Each
Quantity
Catalog number: L19869.06
also known as L19869-06
Price (USD)
Price: 212.00
Online price: 180.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6937-03-7
IUPAC Name
methyl 2-aminopyridine-4-carboxylate
Molecular Formula
C7H8N2O2
InChI Key
SVWWNEYBEFASMP-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=NC(N)=C1
Appearance (Color)
Pale green or pale cream to brown
Melting Point
143-156°C
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%

Description

Methyl 2-aminopyridine-4-carboxylate is an inhibitor of nitrogen monoxide synthetase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-aminopyridine-4-carboxylate is an inhibitor of nitrogen monoxide synthetase.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text