1-Decylboronic acid, 98%
1-Decylboronic acid, 98%
1-Decylboronic acid, 98%
Thermo Scientific Chemicals

1-Decylboronic acid, 98%

CAS: 24464-63-9 | C10H23BO2 | 186.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L19957.03
also known as L19957-03
Price (EUR)
35,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
35,00
Each
Chemical Identifiers
CAS24464-63-9
IUPAC Namedecylboronic acid
Molecular FormulaC10H23BO2
InChI KeyTUTSEMMBKGBUGD-UHFFFAOYSA-N
SMILESCCCCCCCCCCB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale cream
Proton NMRConforms to structure
Assay (GC)≥92.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only