2-Bromo-3-nitropyridine, 98%
2-Bromo-3-nitropyridine, 98%
2-Bromo-3-nitropyridine, 98%
Thermo Scientific Chemicals

2-Bromo-3-nitropyridine, 98%

CAS: 19755-53-4 | C5H3BrN2O2 | 203.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20019.03
also known as L20019-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS19755-53-4
IUPAC Name2-bromo-3-nitropyridine
Molecular FormulaC5H3BrN2O2
InChI KeyZCCUFLITCDHRMG-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CN=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)120.0-126.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only