3H-1,2-Benzodithiol-one 1,1-dioxide, 98%
3H-1,2-Benzodithiol-one 1,1-dioxide, 98%
3H-1,2-Benzodithiol-one 1,1-dioxide, 98%
Thermo Scientific Chemicals

3H-1,2-Benzodithiol-one 1,1-dioxide, 98%

CAS: 66304-01-6 | C7H4O3S2 | 200.226 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 L20129.03
亦稱為 L20129-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS66304-01-6
IUPAC Name3H-1λ⁶,2-benzodithiole-1,1,3-trione
Molecular FormulaC7H4O3S2
InChI KeyJUDOLRSMWHVKGX-UHFFFAOYSA-N
SMILESO=C1SS(=O)(=O)C2=CC=CC=C12
檢視更多
規格Specification Sheet規格表
Identification (FTIR)Conforms
Appearance (Color)Colorless to white
Water Content (Karl Fischer Titration)≤0.5%
Melting Point (clear melt)100-107?C
Assay (HPLC)≥97.5%
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only