1-Bromo-4-tert-butoxybenzene, 98%
Thermo Scientific Chemicals

1-Bromo-4-tert-butoxybenzene, 98%

Have Questions?
Catalog NumberQuantity
L20158.03
also known as L20158-03
1 g
Catalog number L20158.03
also known as L20158-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS60876-70-2
IUPAC Name1-bromo-4-(tert-butoxy)benzene
Molecular FormulaC10H13BrO
InChI KeyQIWQHUCUWNGYDZ-UHFFFAOYSA-N
SMILESCC(C)(C)OC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or Low melting solid
Appearance (Color)White to pale yellow.
Assay (GC)≥97.5%
Melting Point (clear melt)32-37?C
It is used in the stereoselective synthesis of four stereoisomers of β-Methoxytyrosine, a component of callipeltin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.