4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis
4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis
4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis
Thermo Scientific Chemicals

4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis

CAS: 16208-17-6 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20274.03
also known as L20274-03
Price (EUR)
70,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
70,70
Each
Chemical Identifiers
CAS16208-17-6
IUPAC Name2,2-dihydroxy-1-(4-methoxyphenyl)ethan-1-one
Molecular FormulaC9H10O4
InChI KeyNLUFPYPVEHQAQN-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(=O)C(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Water Content (Karl Fischer Titration)<28.0%
Appearance (Color)White to yellow
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only