Thermo Scientific Chemicals

2-Chloropyridine-3-boronic acid, 96%, Thermo Scientific Chemicals

Catalog number: L20303.03
1 g, Each
Thermo Scientific Chemicals

2-Chloropyridine-3-boronic acid, 96%, Thermo Scientific Chemicals

Catalog number: L20303.03
1 g, Each
Quantity
Catalog number: L20303.03
also known as L20303-03
Price (USD)
Price: 95.10
Online price: 81.65
Your price:
Quantity
-

Chemical Identifiers

CAS
381248-04-0
IUPAC Name
(2-chloropyridin-3-yl)boronic acid
Molecular Formula
C5H5BClNO2
InChI Key
VRDAOVQZVXYRNH-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CN=C1Cl
Proton NMR
Conforms to structure
Assay (Aqueous acid-base Titration)
> 95.0%
Form
Powder
Assay (HPLC)
> 95.0%
Appearance (Color)
White to cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text