1-Heptadecanol
1-Heptadecanol
1-Heptadecanol
Thermo Scientific Chemicals

1-Heptadecanol

CAS: 1454-85-9 | C17H36O | 256.474 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L20404.14
also known as L20404-14
Price (EUR)
246,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
246,00
Each
Chemical Identifiers
CAS1454-85-9
IUPAC Nameheptadecan-1-ol
Molecular FormulaC17H36O
InChI KeyGOQYKNQRPGWPLP-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Formcrystalline or fused solid
Assay (GC)≥ 95.0%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only