Thermo Scientific Chemicals

2-Ethoxypyridine-3-boronic acid, 98%, Thermo Scientific Chemicals

Catalog number: L20422.03
1 g, Each
Thermo Scientific Chemicals

2-Ethoxypyridine-3-boronic acid, 98%, Thermo Scientific Chemicals

Catalog number: L20422.03
1 g, Each
Quantity
Catalog number: L20422.03
also known as L20422-03
Price (USD)
Price: 182.00
Online price: 157.65
Your price:
Quantity
-

Chemical Identifiers

CAS
854373-97-0
IUPAC Name
(2-ethoxypyridin-3-yl)boronic acid
Molecular Formula
C7H10BNO3
InChI Key
FXUMKSCYKPOZOO-UHFFFAOYSA-N
SMILES
CCOC1=NC=CC=C1B(O)O
Appearance (Color)
White to pale brown
Assay (Aqueous acid-base Titration)
≥97.5%
Melting Point (clear melt)
101.5-107.5?C
Assay (HPLC)
≥97.5%
Proton NMR
Conforms to structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text