Lipopolysaccharides From Escherichia coli Serotype 055:B5, Alexa Fluor™ 594 Conjugate
This bright, red-fluorescent Alexa Fluor 594 conjugate can be used to follow lipopolysaccharide binding, transport and cell-internalization processes.Read more
Have Questions?
Catalog Number
Quantity
L23353
also known as L-23353
100 μg
Catalog number L23353
also known as L-23353
Price (USD)
317.00
Each
Quantity:
100 μg
Price (USD)
317.00
Each
This bright, red-fluorescent Alexa Fluor 594 conjugate can be used to follow lipopolysaccharide binding, transport and cell-internalization processes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeAlexa Fluor Dyes
Excitation/Emission590/617
FormSolid
Quantity100 μg
Shipping ConditionRoom Temperature
Product LineAlexa Fluor
Product TypeLipopolysaccharide Conjugate
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Citations & References (4)
Citations & References
Abstract
Inhibition of Cholesteryl Ester Transfer Protein Preserves High-Density Lipoprotein Cholesterol and Improves Survival in Sepsis.
Authors:
Journal:Circulation
PubMed ID:33228395
Brain-localized and Intravenous Microinjections in the Larval Zebrafish to Assess Innate Immune Response.
Authors:
Journal:Bio Protoc
PubMed ID:33889672
A Macrophage-Magnesium Hybrid Biomotor: Fabrication and Characterization.
Authors:
Journal:Adv Mater
PubMed ID:31070278
Advanced glycation end-products reduce lipopolysaccharide uptake by macrophages.