Product Image
Invitrogen™

LI Silver (LIS) Enhancement Kit

The LI Silver (LIS) Enhancement Kit is a convenient, light-insensitive silver enhancement system for use with our colloidal gold andRead more
Have Questions?
Catalog NumberQuantity
L249191 kit
Catalog number L24919
Price (USD)
314.65
Online exclusive
324.00
Save 9.35
Each
Quantity:
1 kit
Price (USD)
314.65
Online exclusive
324.00
Save 9.35
Each
The LI Silver (LIS) Enhancement Kit is a convenient, light-insensitive silver enhancement system for use with our colloidal gold and NANOGOLD reagents. Gold particles in the presence of silver (I) ions and a reducing agent act as catalysts to reduce the silver (I) ions to metallic silver. The metallic silver is deposited onto the gold, enlarging the particles to between 30 and 100 nm in diameter. Tissue or blots stained with colloidal gold are “developed” by this autometallographic procedure to give black staining that can be seen in the light microscope. The LI Silver Enhancement Kit is prepared for Molecular Probes by Nanoprobes, Inc.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeNANOGOLD™, FluoroNanogold™, Silver, Colloidal Gold
Product TypeLIS Enhancement Kit
Quantity1 kit
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Citations & References (23)

Citations & References
Abstract
Immunolocalization of a Drosophila phosphatidylinositol transfer protein (rdgB) in normal and rdgA mutant photoreceptor cells with special reference to the subrhabdomeric cisternae.
Authors:Suzuki E, Hirosawa K
Journal:J Electron Microsc (Tokyo)
PubMed ID:7996077
'Distribution of rdgB protein, which was recently shown to be a Drosophila phosphatidylinositol transfer protein, was studied in the photoreceptor cells of compound eyes of normal and rdgAPC47 mutant of Drosophila melanogaster by immunoelectron microscopy using (1) pre-embedding HRP staining, (2) pre-embedding NANOGOLD labeling followed by siliver enhancement, (3) and ... More
A practical technique to postfix nanogold-immunolabeled specimens with osmium and to embed them in Epon for electron microscopy.
Authors:Sawada H, Esaki M
Journal:J Histochem Cytochem
PubMed ID:10727291
'Nanogold is a tiny gold probe, freely diffusible in cells and tissues, and is suitable for pre-embedding immunohistochemistry. However, it is necessary to develop Nanogold to a larger size so that it can be observed by conventional transmission electron microscopy. Silver enhancement is usually used for visualizing Nanogold, but the ... More
Synaptotagmin VII as a plasma membrane Ca(2+) sensor in exocytosis.
Authors:Sugita S, Han W, Butz S, Liu X, Fernández-Chacón R, Lao Y, Südhof TC
Journal:Neuron
PubMed ID:11395007
'Synaptotagmins I and II are Ca(2+) binding proteins of synaptic vesicles essential for fast Ca(2+)-triggered neurotransmitter release. However, central synapses and neuroendocrine cells lacking these synaptotagmins still exhibit Ca(2+)-evoked exocytosis. We now propose that synaptotagmin VII functions as a plasma membrane Ca(2+) sensor in synaptic exocytosis complementary to vesicular synaptotagmins. ... More
Cryo-electron microscopic localization of protein L7/L12 within the Escherichia coli 70 S ribosome by difference mapping and Nanogold labeling.
Authors:Montesano-Roditis L, Glitz DG, Traut RR, Stewart PL
Journal:J Biol Chem
PubMed ID:11278411
'The Escherichia coli ribosomal protein L7/L12 is central to the translocation step of translation, and it is known to be flexible under some conditions. The assignment of electron density to L7/L12 was not possible in the recent 2.4 A resolution x-ray crystallographic structure (Ban, N., Nissen, P., Hansen, J., Moore, ... More
Highly sensitive DNA, RNA and antigen detection methods: streptavidin-Nanogold-silver staining.
Authors:Hauser-Kronberger C
Journal:Cell Vis
PubMed ID:9660736