Lectin PNA From <i>Arachis hypogaea </i>(peanut), Alexa Fluor&trade; 647 Conjugate
Invitrogen™

Lectin PNA From Arachis hypogaea (peanut), Alexa Fluor™ 647 Conjugate

Lectin PNA is specific for terminal β-galactose. It will agglutinate human erythrocytes, but only after neuraminidase treatment.View complete list of자세히 알아보기
Have Questions?
카탈로그 번호수량
L324601 mg
카탈로그 번호 L32460
제품 가격(KRW)
301,000
온라인 행사
Ends: 30-Sep-2026
343,000
할인액 42,000 (12%)
Each
수량:
1 mg
제품 가격(KRW)
301,000
온라인 행사
Ends: 30-Sep-2026
343,000
할인액 42,000 (12%)
Each
Lectin PNA is specific for terminal β-galactose. It will agglutinate human erythrocytes, but only after neuraminidase treatment.

View complete list of fluorescent dye-conjugated lectins ›

For Research Use Only. Not for use in diagnostic procedures.
사양
라벨 유형Alexa Fluor Dyes
제품라인Alexa Fluor
단백질 아형Lectins
수량1 mg
배송 조건Room Temperature
소스Peanut
콘주게이트Alexa Fluor 647
형태Lyophilized
Unit SizeEach
구성 및 보관
Store in freezer (-5 to -30°C) and protect from light.

인용 및 참조 문헌 (10)

인용 및 참조 문헌
Abstract
ΔN-Tp63 Mediates Wnt/β-Catenin-Induced Inhibition of Differentiation in Basal Stem Cells of Mucociliary Epithelia.
Authors:
Journal:Cell Rep
PubMed ID:31553905
Usher syndrome type 1-associated cadherins shape the photoreceptor outer segment.
Authors:
Journal:J Cell Biol
PubMed ID:28495838
The Deubiquitinating Enzyme Ataxin-3 Regulates Ciliogenesis and Phagocytosis in the Retina.
Authors:
Journal:Cell Rep
PubMed ID:33176149
Mature Retina Compensates Functionally for Partial Loss of Rod Photoreceptors.
Authors:
Journal:Cell Rep
PubMed ID:32521255
Increasing Ca2+ in photoreceptor mitochondria alters metabolites, accelerates photoresponse recovery, and reveals adaptations to mitochondrial stress.
Authors:
Journal:Cell Death Differ
PubMed ID:31371786