LIVE/DEAD™ Fixable Yellow Dead Cell Stain Kit, for 405 nm excitation
Invitrogen™

LIVE/DEAD™ Fixable Yellow Dead Cell Stain Kit, for 405 nm excitation

The LIVE/DEAD™ Fixable Yellow Dead Cell Stain Kit is used to determine the viability of cells prior to the fixationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
L34968400 Assays
L34959200 Assays
L34967
also known as L-34967
80 assays
Catalog number L34968
Price (USD)
436.00
Each
Quantity:
400 Assays
Price (USD)
436.00
Each
The LIVE/DEAD™ Fixable Yellow Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation. This kit has been optimized and validated for use with a violet laser flow cytometer.

• Stable—dyes are freeze dried in separate vials to maintain stability

• Robust—staining pattern is the same before and after fixation

• Low compensation—minimal spectral overlap between other fluorophores

View a selection guide for all fixable viability dyes for flow cytometry.

Stable
Unlike products that are sold in solution, the LIVE/DEAD™ Fixable Yellow Stain has been conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time. Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated. If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles.

Robust
Dead cell discriminator stains can lose sensitivity after treatment with fixatives such as formaldehyde or ethanol-based fixation methods required for intracellular phosphorylation studies. The LIVE/DEAD™ Fixable Yellow Stain is an amine reactive dye that binds covalently to intracellular and extracellular amines, and the staining pattern is preserved following formaldehyde fixation.

Low compensation
The LIVE/DEAD™ Fixable Yellow Stain was selected based on its fluorescent properties to minimize compensation between other violet dyes and dyes that excite off of the 488 nm blue laser. The yellow-fluorescent reactive dye has an excitation maximum of ∼405 nm, but it is excited well with the 405 nm violet laser, and it has an emission maxima of ∼570 nm, so it can be collected in the second or third channel on most violet laser flow cytometers.

How it works
In cells with compromised membranes, the dye reacts with free amines both in the cell interior and on the cell surface, yielding intense fluorescent staining. In viable cells, the dye's reactivity is restricted to the cell-surface amines, resulting in less intense fluorescence. The difference in intensity is typically greater than 50-fold between live and dead cells, allowing for easy discrimination.

Colors available
LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet your multi-color panel needs.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityImpermeant
Cell TypeEukaryotic Cells
DescriptionLIVE/DEAD™ Fixable Yellow Dead Cell Stain Kit, for 405 nm excitation
Detection MethodFluorescence
Dye TypeLIVE/DEAD™ Fixable Yellow Dead Cell Stain
Emission Wavelength570
Excitation Wavelength405 nm
FormSolid
FormatTube(s)
Quantity400 Assays
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
ColorYellow
For Use With (Application)Viability Assay
For Use With (Equipment)Flow Cytometer
Product LineLIVE/DEAD
Product TypeStain
Unit SizeEach
Contents & Storage
Contains 10 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO.

Store at -20°C.

Frequently asked questions (FAQs)

How long can I store LIVE/DEAD Fixable Yellow Dead Cell Stain Kit, for 405 nm excitation (Cat. No. L34959) at -20 degrees C?

Once reconstituted, the Live/Dead fixable dyes are guaranteed to be stable at -20 degrees C for a few weeks. We have not tested storage conditions beyond this time frame. The dye breaks down in water so the condition of the DMSO that is used for resuspension is the most important determinant.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How do I prepare dead cell controls for LIVE/DEAD cell viability assays?

There are two easy options. One is to heat-inactivate the cells by placing at 60 degrees C for 20 minutes. The second is to subject the cells to 70% ethanol. Alcohol-fixed cells can be stored indefinitely in the freezer until use, potentially up to several years.

Centrifuge cells, pellet, and remove supernatant.
Fix cells: Add 10 mL ice cold 70% ETOH to a 15 mL tube containing the cell pellet, adding dropwise at first while vortexing, mix well.
Store in freezer until use.
When ready to use, wash twice and resuspend in buffer of choice.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Which cell viability kits are compatible with fixation?

The LIVE/DEAD Fixable kits for flow cytometry analysis are compatible with fixation. These kits use amine-reactive cell-impermeant dyes that stain the cell surface of live cells and also the cytosol of dead cells-live cells are dim and dead cells are bright. Since the dye is covalently bound to the cells, it will be retained after fixation. Unfortunately, this method does not work well for imaging-based assays, as all cells are stained and it is difficult to distinguish bright dead cells from dim live cells with a microscope. Ethidium monoazide (EMA; Cat No. E1374) is a cell impermeant nucleic acid stain that can be applied to live cultures and stains only dead cells. After incubation and washing away unbound dye, the cells can be exposed to light to photoactivate EMA to crosslink to dead cell DNA. After crosslinking to dead cell DNA, the samples may be fixed and permeabilized. Image-IT DEAD Green Viability Stain (Cat. No. I10291) for imaging and high-content screening (HCS) analysis is a live-cell impermeant DNA binding dye that is compatible with fixation and permeabilization with good retention up to 48 hours. We also have a LIVE/DEAD Reduced Biohazard Cell Viability Kit (Cat. No. L7013) for imaging and flow analysis that contains two DNA binding dyes, SYTO 10 and Dead Red, that are sufficiently retained to be analyzed soon after 4% glutaraldehyde fixation.
Note: In general, DNA-binding dyes and calcein AM are not compatible with fixation, as these dyes are not covalently bound to components of the cell and will thus slowly diffuse out of cells after fixation, gradually staining all cells as dead.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.