LIVE/DEAD™ Fixable Lime (506) Viability Kit, for 405 nm excitation, 400 Assays - FAQs
LIVE/DEAD™ Fixable Lime (506) Viability Kit, for 405 nm excitation, 400 Assays - FAQs
View additional product information for LIVE/DEAD™ Fixable Lime (506) Viability Kit, for 405 nm excitation - FAQs (L34991, L34990, L34989)
1 product FAQs found
What is the concentration of the LIVE/DEAD Fixable dyes?
The concentration of LIVE/DEAD Fixable dyes is proprietary.