LysoLight™ Antibody Labeling Kits
Invitrogen™
LysoLight™ Antibody Labeling Kits
The LysoLight Antibody Labeling Kits offer an efficient method for conjugating antibodies with LysoLight dyes. This process allows for specific and sensitive detection of internalized antibodies within lysosomes using fluorescence microscopy, flow cytometry, or high content imaging platforms.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadTipo de productoColor
L36004500 μgAmine Reactive DyeDeep Red
L360013 x 100 μgAntibody Labeling KitDeep Red
L360023 x 1 mgAntibody Labeling KitDeep Red
L360033 x 100 μgAmine Reactive DyeDeep Red
Número de catálogo L36004
Precio (MXN)
-
Cantidad:
500 μg
Tipo de producto:
Amine Reactive Dye
Color:
Deep Red
LysoLight Antibody Labeling Kits provide a powerful tool for visualizing the lysosomal localization and catabolic degradation of antibodies and proteins. These kits utilize a unique fluorophore that is only active upon enzymatic cleavage by the lysosomal specific enzyme, Cathepsin B. Outside of the lysosome, LysoLight dyes have little to no observable background fluorescence, and only upon localization to the protease rich environment of the lysosome, does LysoLight become activated and brightly fluoresce bright green or deep red in color.
The labeling protocol provided in the LysoLight Antibody Labeling Kits involves a simple and rapid procedure, ensuring minimal hands-on time and reliable results. The kits include all necessary reagents, including the LysoLight dye, buffers, and purification columns, to facilitate the antibody labeling process. The LysoLight reactive dye contained in these kits readily reacts with lysine residues in antibodies or proteins to yield a covalently attached fluorogenic sensor. The spin columns included in the kits are used for purifying the labeled antibody from excess dye with yields of 70–95%. The kits contain sufficient reagents for three labeling reactions of 100 μg or 1 mg of antibody. The user guide describes a general protocol for using the amine-reactive form of the LysoLight dye for labeling either 100 μg or 1 mg of IgG.
Para uso exclusivo en investigación.
Especificaciones
DescripciónLysoLight™ Deep Red Amine Reactive Dye
Método de detecciónFluorescente
Excitación/emisión650/668 nm
Cantidad500 μg
SolubilidadDMSO (dimetilsulfóxido)
ColorDeep Red
Para utilizar con (aplicación)Antibody and Protein Labeling
Para utilizar con (equipo)Flow Cytometer, Fluorescence Microscope, High Content Analysis Instrument
Línea de productosLysoLight
Tipo de productoAmine Reactive Dye
Unit SizeEach