Novex™ Zymogram Renaturing Buffer (10X) - FAQs
Novex™ Zymogram Renaturing Buffer (10X) - FAQs
View additional product information for Novex™ Zymogram Renaturing Buffer (10X) - FAQs (LC2670)
1 product FAQs found
Does the Colloidal Blue Staining kit work with Invitrogen Zymogram gels?
Yes, the Colloidal Blue Staining kit works very well with Invitrogen Zymogram gels.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.