Novex™ Tris-Glycine Native Sample Buffer (2X) - FAQs
Novex™ Tris-Glycine Native Sample Buffer (2X) - FAQs
View additional product information for Novex™ Tris-Glycine Native Sample Buffer (2X) - FAQs (LC2673)
1 product FAQs found
Where do I find buffer recipes for your precast protein gels?
The formulations of buffers for our precast protein gels can be found at this link: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-electrophoresis-buffers-reagents.html
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.