Novex™ Rapid Tris-Glycine SDS Running Buffer (Powder)
Invitrogen™

Novex™ Rapid Tris-Glycine SDS Running Buffer (Powder)

Invitrogen Novex Rapid Tris-Glycine SDS Running Buffer (Powder) delivers more than 50% faster SDS-PAGE run times compared to traditional Tris-Glycine buffers, without compromising separation performance. Compatible with all Tris-Glycine gels, it offers room temperature storage and a 2-year shelf life.
Have Questions?
Catalog NumberQuantity
LC26785 Packets
Catalog number LC2678
Price (MXN)
-
Quantity:
5 Packets

Invitrogen Novex Rapid Tris-Glycine SDS Running Buffer (Powder) is designed to significantly reduce SDS-PAGE run times while maintaining separation performance. By enabling separations over 50% faster than traditional Tris-Glycine running buffers, this rapid buffer supports more efficient protein electrophoresis without compromising band resolution. Compatible with all Tris-Glycine gels, including both precast and handmade formats, the powder form offers flexible preparation, room temperature storage, a two-year shelf life, and is designed for convenient routine use.

Features and benefits include:

  • More than 50% faster SDS-PAGE separations—significantly reduces electrophoresis run times compared to traditional Tris-Glycine running buffers, accelerating protein analysis workflows
  • Maintains separation performance—faster run times achieved with minimal impact to band resolution and overall separation quality
  • Compatible with all Tris-Glycine gels—designed for use with Tris-Glycine gel systems, including both Novex Tris-Glycine and SureCast handcast gels
  • Powder format for flexible preparation—prepare fresh running buffer as needed to support consistent electrophoresis performance and simplify laboratory workflows
  • Room temperature storage and long shelf life—powder format enables convenient storage with a two-year shelf life for improved inventory management
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)SDS-PAGE
For Use With (Equipment)Electrophoresis Chambers
FormulationRapid Tris-Glycine SDS Running Buffer
Quantity5 Packets
Shelf Life2 Years
Storage RequirementsRoom Temperature
Sufficient ForPreparing 5 L Running Buffer
ColorWhite
FormatPowder
Gel TypeTris-Glycine
Product LineNovex
Product TypeSDS Running Buffer
Unit SizeEach