Novex™ Rapid Tris-Glycine SDS Running Buffer (10X, 500mL)
Invitrogen™

Novex™ Rapid Tris-Glycine SDS Running Buffer (10X, 500mL)

Invitrogen Novex Rapid Tris-Glycine SDS Running Buffer (10X, 500 mL) delivers over 50% faster SDS-PAGE run times than traditional Tris-Glycine buffers without compromising separation performance. Supplied as a ready-to-use liquid with a 12-month shelf life, stored at 2°C to 8°C.
Have Questions?
Catalog NumberQuantity
LC26795 Packets
Catalog number LC2679
Price (USD)
45.00
Each
Quantity:
5 Packets
Price (USD)
45.00
Each

Invitrogen Novex Rapid Tris-Glycine SDS Running Buffer (10X, 500 mL) is designed to significantly reduce SDS-PAGE run times while maintaining separation performance. By enabling separations over 50% faster than traditional Tris-Glycine running buffers, this rapid buffer supports more efficient protein electrophoresis without compromising band resolution. Supplied as a ready-to-use liquid concentrate, the 10X format allows convenient dilution and consistent preparation, with a 12-month shelf life when stored refrigerated at 2–8°C.

Features and benefits include:

  • More than 50% faster SDS-PAGE separations—significantly reduces electrophoresis run times compared to traditional Tris-Glycine running buffers, accelerating protein analysis workflows
  • Maintains separation performance—faster run times achieved with minimal impact to band resolution and overall separation quality
  • Compatible with all Tris-Glycine gels—designed for use with Tris-Glycine gel systems, including both Novex Tris-Glycine and SureCast handcast gels
  • Concentrated 10X format—ready-to-use liquid allows simple dilution and consistent buffer preparation
  • Refrigerated storage with defined shelf life—liquid format requires storage at 2–8°C and offers a 12-month shelf life for reliable routine use
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)SDS-PAGE
For Use With (Equipment)Electrophoresis Chambers
FormulationRapid Tris-Glycine SDS Running Buffer
Quantity5 Packets
Shelf Life365 Days
Storage RequirementsStore at 2°C to 8°C.
Sufficient ForPreparing 5 L Running Buffer
Volume (Metric)500 mL
ColorClear
FormatBottle
Gel TypeTris-Glycine
Product LineNovex
Product TypeSDS Running Buffer
Unit SizeEach