E-PAGE™ MagicMark™ Unstained Protein Standard

Catalog number: LC5701

Novex™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The E-PAGE™ SeeBlue™ Pre-stained and MagicMark™ Unstained Protein Standards are designed for use with the pre-cast E-PAGE™ Electrophoresis System:

• The E-PAGE™ SeeBlue® Pre-stained Standard consists of three blue-dyed and two colored bands ranging in size from 21-261 kDa and is suitable for monitoring gel and transfer runs.
• The E-PAGE™ MagicMark™ Unstained Protein Standard consists of five recombinant proteins ranging in size from 20-220 kDa, each with an IgG binding-site. The E-PAGE™ MagicMark™ Standard proteins can be detected with Coomassie® stain on the gels or conjugated-antibody directly on western blots. It is suitable for molecular weight estimation of unknown proteins.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Compatibility: E-PAGE™
Product Line: E-PAGE™, MagicMark™
Product Size: 250 µL
Ready to Load: Yes
Size Range: 20 to 220 kDa
Stain Type: Unstained
System: E-PAGE™
Shipping Condition: Dry Ice

Contents & storage

500 µl (50 applications of 10 µl each) of the E-PAGE™ SeeBlue® Pre-stained Standard is provided in loading buffer consisting of Tris-HCl, Formamide, and SDS. Store at +4°C. 250 µl (50 applications of 5 µl each) of the E-PAGE™ MagicMark™ Unstained Protein Standard is provided in loading buffer consisting of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 1% SDS, and 0.025% Bromophenol Blue. Store at -20°C. Avoid repeated freezing and thawing.

Documents

Manuals & protocols

Recommended products