Filter Plate

Catalog number: LCP0000

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Luminex 96-well Filter Plate.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Luminex® Instruments
Format: 96-well plate
Plate Type: Filter Plate
Product Line: Luminex™
Product Size: 1 plate
Shipping Condition: Room Temperature

Contents & storage

Contains 1 microwell filter plate and 4 plate seals.

Documents

Manuals & protocols