Search
| Número de catálogo | Cantidad |
|---|---|
| LF0100A | 45 tiras reactivas |
| LF0200A | 50 tiras reactivas |
Thermo Scientific™ PrecisCheck™ Listeria Kits each contain 50 test strips that utilize advanced lateral flow immunology to produce a positive test line in the presence of the target organism. Use to confirm presumptive-positive colonies gown on Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) as part of the Thermo Scientific™ Listeria Precis™ Methods. Listeria species appear as blue colonies, and L. monocytogenes as blue colonies with an opaque white halo, when grown on Brilliance Listeria Agar (ISO). These presumptive-positive colonies can be confirmed directly from the plate using a test strip from the respective Precis Check Kit.
Rapidly confirm presumptive-positive colonies of Listeria species and Listeria monocytogenes in as little as 15 minutes with the Precis Check Listeria species and Precis Check L. monocytogenes Kits.