Human Ultrasensitive Buffer Reagent Kit

Catalog number: LHB0003M

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Human Ultrasensitive Buffer Reagent Kit for the Luminex® platform is specifically designed to be used with the Human Ultrasensitive Magnetic Bead kits. This buffer kit includes the following components: wash solution, standard reconstitution buffer, assay diluents, incubation buffer, biotin diluents, streptavidin-RPE concentrate and diluents, a 96-well flat bottom plate and a black lid cover. This buffer kit is sufficient for running a full 96-well plate. It can be used with a single Human Ultrasensitive Magnetic Bead kit or with multiple bead kits for multiplexing purposes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Luminex®
Product Size: 100 tests
Species: Human
Shipping Condition: Wet Ice

Contents & storage

Incubation buffer, standard reconstitution buffer, assay diluent, biotin-diluent, SAV concentrate, SAV diluent, wash solution, flat-bottom plate, black lid cover, and protocol. Store at 2-8°C.

Documents

Manuals & protocols

Recommended products