規格
Capacity (Metric)1000 mL
Film DescriptionAegis5-14
For Use With (Application)Bioprocessing
Latex FreeYes
Port Detail3 ports
Product LineLabtainer™
Quantity1 Ea.
Sterilization MethodGamma Irradiation
Tubing Length Line 112 in.
Tubing Length Line 212 in.
Tubing Length Line 34 in.
Tubing Material Line 1MPC body, TPE Tubing, 3/8 in. ID
Tubing Material Line 2MPC insert, TPE Tubing, 3/8 in. ID
Tubing Material Line 3Luer lock body with needleless Luer insert, TPE Tubing, 1/8 in. ID
Tubing StyleTPE
AutoclavableNot Autoclavable
Closure TypeClosed System
ColorTransparent
DisposableSingle-use
MaterialPolyethylene
Product TypeBioprocess Container for Liquid
SterilitySterile
Unit SizeEach