TRACE™ 1310 GC Analyzers for Oxygenates
Thermo Scientific™
TRACE™ 1310 GC Analyzers for Oxygenates
Analyze refinery product streams for oxygenates and octane enhancers according to ASTM methods using Thermo Scientific™ TRACE™ 1310 GC Analyzers for Oxygenates.
Have Questions?
Catalog NumberFunction
LLVLOX0010112Trace Oxygenates in Spark-ignition Engine Fuel (ASTM D7754)
Catalog number LLVLOX0010112
Price (MXN)
-
Function:
Trace Oxygenates in Spark-ignition Engine Fuel (ASTM D7754)
Automatically analyze single or multiple samples using the Thermo Scientific™ TRACE™ 1310 Analyzers for Oxygenates. These automated laboratory gas chromatography systems measure and report oxygenates and octane enhancers in refinery product streams. Systems are preconfigured, method tested, and ready to analyze samples after installation. These systems can use the powerful Thermo Scientific™ Chromeleon™ Chromatotraphy Data System (CDS) to acquire and analyze chromatography data. Custom reporting allows generation of customer-specific reports. Each system is delivered with methods pre-installed and a complete documentation package that includes start-up, troubleshooting, plumbing diagram, and spare parts/consumables list.
We offer three systems configured for specific hydrocarbon streams and ASTM standard test methods. All systems utilize automated backflush of stripper column to prevent contamination of sensitive oxygentates column thus increasing uptime and performance.
TRACE™ 1310 GC for Oxygenates by ASTM D7423
  • FID Channel detects and quantitates oxygenate compounds in light hydrocarbon streams C2-C5 as specified in ASTM D7423. Stripper column holds hydrocarbons up for backflush to prevent contamination of oxygenates column.
TRACE™ 1310 GC for Oxygenates by ASTM D7754
  • FID Channel detects and quantitates oxygenate compounds in spark ignition fuels (gasoline) as specified in the ASTM D7754. Stripper column holds hydrocarbons which are vented by a pressure point tee to prevent contamination of oxygenates column.
TRACE™ 1310 GC for Oxygenates in Gasoline by ASTM D4815
  • FID Channel detects and quantitates oxygenate compounds in spark ignition fuels (gasoline) as specified in the ASTM D4815. Stripper column holds hydrocarbons which are vented by a pressure point Tee to prevent contamination of oxygenates column.
Built to maximize productivity and uptime
  • Systems features Instant Connect detector modules increasing uptime with detector exchange in minutes
  • Heated sample bulkheads ensures complete sample transfer
  • Automated injection for liquid samples
  • Optimized system footprint to reduce benchspace requirement
Specifications
DescriptionGas Chromatography Systems
Detection ModeFlame Ionization
FunctionTrace Oxygenates in Spark-ignition Engine Fuel (ASTM D7754)
IncludesGas chromatograph, inlet, FID, columns and method
Detector TypeFID
Product TypeGC Analyzer for Oxygenates
Unit SizeEach