MAS™ LabLink xL DAILY MODULE
Thermo Scientific™
MAS™ LabLink xL DAILY MODULE
Perform web-based routine QC analysis for day-to-day performance verification with the MAS LabLink xL DAILY MODULE application.
Have Questions?
Catalog NumberSample Type
LLXLDMVarious
Catalog number LLXLDM
Price (USD)
-
Sample Type:
Various
Perform web-based routine QC analysis for day-to-day performance verification with the Thermo Scientific™ MAS™ LabLink xL DAILY MODULE application. Together with Thermo Scientific™ MAS™ LabLink xL WEB, the LabLink xL DAILY MODULE allows laboratories to conduct both internal QC analysis and external real-time peer comparison. QC results are evaluated immediately against defined laboratory criteria. QC data can be manually entered or uploaded electronically via the Thermo Scientific™ MAS™ LabLink xL COMMUNICATOR.
  • 100% web-based application does daily QC comparison
  • Laboratory QC performance tracked using Westgard rule criteria
  • Integrated module within LabLink xL WEB—no software to install
  • LabLink reports provide enhanced internal and external QC comparison
  • An interactive Levey-Jennings plotting tool provides a single screen for primary review and approval
  • Manual or electronic QC results entry—data is immediately reviewed against laboratory defined criteria
  • 100% real-time data processing and report production
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionLabLink xL DAILY MODULE
TypeQC Analytics
Sample TypeVarious
TechnologyQC Analytics
Unit SizeEach