Sterile Automation Tips for Tecan™ Liquid Handling Platforms, 1000μL, clear, filtered
Thermo Scientific™
Sterile Automation Tips for Tecan™ Liquid Handling Platforms, 1000μL, clear, filtered
Increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Have Questions?
Número do catálogoVolume (Metric)
LMF175-96RS10001000 μL
Número do catálogo LMF175-96RS1000
Preço (BRL)
-
Volume (Metric):
1000 μL
Increase pipetting performance with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
  • Sterile, filtered tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
Especificações
ColorNatural
FilteredFiltered
For Use With (Equipment)Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro
FormatRack
Product LineTecan
SterilitySterile
Tip StyleAutomation
Volume (Metric)1000 μL
Unit SizeCase of 4800