Oxoid™ Yeast Extract Powder
Thermo Scientific™

Oxoid™ Yeast Extract Powder

Oxoid Yeast Extract Powder is a dried yeast autolysate of proprietary specification optimized for fermentation and culture media applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0021K25 kg
LP0021B500 g
LP0021R2.5 kg
LP0021T5 kg
Catalog number LP0021K
Price (USD)
-
Quantity:
25 kg
Thermo Scientific™ Oxoid Yeast Extract Powder is a proprietary dried yeast autolysate which provides a nutritious source of nitrogen and vitamins optimized for use in culture media or fermentation.

Increases yield of organisms in many media or fermentation broths.

With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
DescriptionYeast Extract Powder
For Use With (Application)Culture media or fermentation processes
Protein SourceYeast Extract
Quantity25 kg
Shelf Life3 years
FormDehydrated Powder
Product TypePeptone
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage