Liver Desiccated Bacteriological
Thermo Scientific™
Liver Desiccated Bacteriological
Prepare liver infusion media with Liver Desiccated Bacteriological.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0026T5 kg
LP0026J250 g
LP0026H12 kg
Catalog number LP0026T
Price (USD)
-
Quantity:
5 kg
Prepare Liver infusion media with Thermo Scientific™ Oxoid™ Liver Desiccated Bacteriological. Liver Desiccated is prepared by the dehydration of fresh pork livers under carefully controlled conditions designed to ensure maximum retention of nutritive properties, and is equivalent to five times its weight of fresh liver
Dehydrated whole liver, specially manufactured for the preparation of infusion media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
AppearanceBrown Granules
ColorBrown
DescriptionLiver Desiccated Bacteriological
Quantity5 kg
Shelf Life5 Years
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
10°C to 30°C