Oxoid™ Peptone Bacteriological
Thermo Scientific™

Oxoid™ Peptone Bacteriological

The Oxoid Peptone Bacteriological is an all-purpose peptone specially prepared for use in fermentation applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0037B500 g
LP0037K25 kg
LP0037T5 kg
Catalog number LP0037B
Price (INR)
-
Quantity:
500 g

Thermo Scientific™ Oxoid Peptone Bacteriological and its neutralized form are very nutritious, all-purpose peptones prepared by the enzymatic digestion of selected animal protein sources. They are specially prepared to be compatible with other refined culture media or fermentation ingredients.

The combination of pancreatin and papain enzyme systems ensures that these bacteriological peptones contain a wide spectrum of polypeptides, reflected in their broad molecular profiles. 

The neutralized form evolved from the original to meet those occasions when a slightly higher pH is required. Both, when reconstituted, give a solution free of haze, cloudiness, or precipitation.

With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
For Use With (Application)Culture media or fermentation processes
Protein SourceBeef
Quantity500 g
Shelf Life5 years
FormDehydrated Powder
Product TypePeptone
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage