Oxoid™ Tryptone T
Thermo Scientific™

Oxoid™ Tryptone T

Oxoid Tryptone T peptone is optimized for low ion levels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0043B500 g
LP0043C400 g
LP0043T5 kg
Catalog number LP0043B
Price (USD)
-
Quantity:
500 g
Thermo Scientific™ Oxoid Tryptone T peptone developed with lower ion levels is ideal for the production of toxin by Clostridium tetani in fermentation applications.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
For Use With (Application)Fermentation processes
Protein SourceCasein
Quantity500 g
Shelf Life720 Days from Manufacture
FormDehydrated Powder
Product TypePeptone
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage