Oxoid™ Lab Lemco No. 2
Thermo Scientific™
Oxoid™ Lab Lemco No. 2
Thermo Scientific™ Oxoid Lab-Lemco Powder is made with premium beef sourced from New Zealand exclusively to deliver a nutritious and safe product.
Have Questions?
Número de catálogoCantidad
LP0129K25 kg
Número de catálogo LP0129K
Precio (MXN)
-
Cantidad:
25 kg
Thermo Scientific™ Oxoid Lab-Lemco Powder is a Beef Extract made from specially selected raw materials for quality and bio-logical safety.
Use with other refined ingredients, e.g. Neutralized bacteriological peptone (LP0034) to prepare culture media which do not require filtration.
May be used in fermentation processes.
LP0129 is manufactured using beef from New Zealand.
Industrial Reference: NMKL
With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.
Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.
Supports quality media design:
  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free
Multi-method suitability:
  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
ColorStraw
Para utilizar con (aplicación)Fermentation processes
Protein SourceBeef
Cantidad25 kg
Duración de almacenamiento720 Days from Manufacture
FormularioDehydrated Powder
Tipo de productoPeptone
Unit SizeEach
Contenido y almacenamiento
Dehydrated Powder, Ambient & Dry Storage