Extra Seat Licenses (Qty 3) - Luminex® 200™ System

Catalog number: LUMEXL

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

xPONENT® 3.1- Extra Seat Licenses (Qty 3), for the Luminex® 200™ system.

Specifications

For Use With (Equipment): Luminex® 200™ System
Product Line: Luminex™
Product Size: 3 seats
Shipping Condition: Room Temperature

Documents

Manuals & protocols