Basic Security Module, Luminex® 200™ System

Catalog number: LX200BS

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The basic security module provides security by allowing an administrator to set up user access levels. This module is designed for the Luminex® 200™ system.

Specifications

For Use With (Equipment): Luminex® 200™ System
Product Line: Luminex™
Product Size: 1 each
Software Type: Security
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols