Invitrogen™

Touch Screen Monitor for Luminex™ System

Catalog number:  LX200TM

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Touch Screen Monitor
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Luminex® Instruments
Product Line: Luminex™
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Monitors

Contents & storage

Store at room temperature.

Documents

Manuals & protocols