Search
| Katalognummer | Zur Verwendung mit (Geräte) |
|---|---|
| LYKOS21CFR | MarqMetrix All-In-One Process Raman-Analyzer |
Attain cGMP and 21 CFR Part 11 FDA compliance when using the Thermo Scientific™ MarqMetrix™ All-In-One Process Raman Analyzer with Thermo Scientific™ Lykos™ PAT Software – Compliance. A remote control and monitoring software that delivers process data like never before, keeping you in the know at every stage of your processes.
Advanced security measures
Detailed auditing
Electronic signatures
Along with the 21 CFR Part 11 software, our service and support team offers IQ/OQ qualification services for your instrument and software.