PEEK Fingertight Fittings
Imagem ilustrativa. O produto pode variar
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
Thermo Scientific™
PEEK Fingertight Fittings
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoTypeDescription
M-645XFingertight FittingPEEK, 1/32 in., 0.25 in. Head
U-467RXColumn End Plugs, Red1/16 in., 10-32 Coned, Delrin, Red
Número do catálogo M-645X
Preço (BRL)
-
Type:
Fingertight Fitting
Description:
PEEK, 1/32 in., 0.25 in. Head
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings. Machined for reliability, these fittings resist cracking, breaking, thread stripping and leaking in both low- and high-pressure applications. Fingertight plumbing ensures ease of use. The fittings are biocompatible for use across a broad range of applications.
Description
  • Resist cracking, breaking, thread stripping and leaking in both low and high pressure applications
  • Biocompatible for a broad range of applications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Especificações
DescriptionPEEK, 1/32 in., 0.25 in. Head
Quantity1 Each
For Use With (Equipment)LC Tubing
MaterialPEEK
TypeFingertight Fitting
Unit SizePack of 10