RevertAid™ Master Mix, with DNase I
RevertAid™ Master Mix, with DNase I
RevertAid™ Master Mix, with DNase I
Thermo Scientific™
RevertAid™ Master Mix, with DNase I
Thermo Scientific RevertAid Master Mix with DNase I is a complete system for efficient synthesis of first strand cDNA from mRNA or total RNA templates.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호반응 수
M16325100 Reactions
M1631550 Reactions
카탈로그 번호 M16325
제품 가격(KRW)
322,000
온라인 행사
Ends: 30-Jun-2026
378,000
할인액 56,000 (15%)
Each
반응 수:
100 Reactions
제품 가격(KRW)
322,000
온라인 행사
Ends: 30-Jun-2026
378,000
할인액 56,000 (15%)
Each
Thermo Scientific RevertAid Master Mix with DNase I is a complete system for efficient synthesis of first strand cDNA from mRNA or total RNA templates. The kit includes recombinant endonuclease (DNase I) to remove contaminating genomic DNA from RNA preps.
For reverse transcription the kit uses RevertAid Reverse Transcriptase (RT), which has lower RNase H activity compared to AMV reverse transcriptase. RevertAid Master Mix provides all cDNA synthesis reaction components in a convenient two-tube master mix. It is optimized for highly efficient cDNA synthesis for two-step quantitative RT-PCR (RT-qPCR) applications. The two-tube formulation reduces reaction time and requires fewer pipetting steps than with the RevertAid RT Reverse Transcription Kit, while maintaining high performance.
Highlights
  • Integrated genomic DNA removal step
  • Fast—reaction time typically 15–30 min
  • Convenient—easy workflow with few pipetting steps
  • Reliable—reverse transcription of both abundant and limited targets
  • Flexible—optimized two-step protocol, enabling multiple PCR reactions from a single reverse transcription reaction
Applications
  • First strand cDNA synthesis for RT-PCR and RT-qPCR
  • Construction of full length cDNA libraries
  • Antisense RNA synthesis
For Research Use Only. Not for use in diagnostic procedures.
사양
최종 제품 유형First-Strand cDNA
반응 수100 Reactions
수량100 Reactions
반응 형식Premixed Components
시약 유형Reverse Transcription
역전사 효소RevertAid
리보뉴클레아제 H 활성Yes
배송 조건Dry Ice
시작 물질RNA
기술Reverse Transcription
용도(애플리케이션)Real Time PCR (qPCR), RT-PCR
반응 속도30 min.
Unit SizeEach
구성 및 보관
2 boxes of RevertAid Master Mix (Cat. No. M1631), each containing:
•20X Enzyme mix (MuLV and RNase inhibitor), 50 μL
•2X RT Buffer Mix (includes dNTPs, random primers, and oligo dT), 500 μL
1 box of DNase I (Cat. No. EN0529), containing:
•DNase I, RNase-free (1 U/μL), 200 μL
•10X Reaction Buffer with MgCl2 for DNase I, 1.25 mL
•50 mM EDTA, 1 mL
-25°C to -15°C