MitoProbe™ TMRM Assay Kit for Flow Cytometry
Thermo Scientific™

MitoProbe™ TMRM Assay Kit for Flow Cytometry

The MitoProbe TMRM Assay Kit contains tetramethylrhodamine methyl ester (TMRM) for the detection of mitochondrial membrane potential state and CCCPRead more
Have Questions?
Catalog NumberQuantity
M20036100 assays
Catalog number M20036
Price (USD)
297.65
Online exclusive
310.00
Save 12.35
Each
Quantity:
100 assays
Price (USD)
297.65
Online exclusive
310.00
Save 12.35
Each
The MitoProbe TMRM Assay Kit contains tetramethylrhodamine methyl ester (TMRM) for the detection of mitochondrial membrane potential state and CCCP for the induction of mitochondrial membrane depolarization, if desired.

• Study mitochondrial membrane potential in live cells
• Quick, easy protocol

Determining the state of mitochondrial health can be achieved by detecting changes in the mitochondrial membrane potential. In a healthy cell with active mitochondria, TMRM is readily sequestered, thereby emitting a red-orange fluorescent signal. When apoptosis is induced or a treatment like CCCP is applied, the mitochondrial membrane is depolarized and the TMRM signal diminishes.

Specifications
Detection MethodFluorescence
Emission Wavelength561
FormatVial(s)
Quantity100 assays
Shipping ConditionWet Ice
For Use With (Application)TMRM Assay
For Use With (Equipment)Flow Cytometer
Product TypeFlow Cytometry Assay Kit
Unit SizeEach
Contents & Storage
  • 1 vial TMRM (100 tests)
  • 1 vial 50 mM CCCP
  • Store at -5°C to -30°C.

Frequently asked questions (FAQs)

In the MitoProbe TMRM Assay Kit for Flow Cytometry, should CCCP be present when adding the TMRM?

Yes. The CCCP should be first added to the cell sample, incubated for 5 mins, and then the TMRM should be added and incubated for another 30 mins.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How does the MitoProbe TMRM Assay Kit for Flow Cytometry (Cat. No.M20036) differ from the Image-iT TMRM Reagent (Cat No. I34361)?

The Image-iT TMRM Reagent is provided as a 100 µM solution of TMRM in DMSO (5 vials, 100 µL in each vial). The MitoProbe TMRM Assay Kit for Flow Cytometry provides the TMRM as a 20 µM stock solution in DMSO (1 vial, 100 µL) and a vial of CCCP. Both kits have the stock solutions as a 1000X concentration, but final working concentrations required for imaging applications are 5-fold higher than what is needed for flow cytometry analysis.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How is TMRM different from the MitoTracker dyes?

TMRM is a dynamic mitochondrial stain. There is no covalent attachment of TMRM to the mitochondria and because of this, it can move in and out of the mitochondria dependent upon changes in the membrane potential. The MitoTracker dyes on the other hand possess a reactive chloromethyl group that allows for their covalent attachment to mitochondria. As a result, once they accumulate within mitochondria that have a normal membrane potential, they are retained.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (4)

Citations & References
Abstract
Mitochondrial Delivery of an Anticancer Drug Via Systemic Administration Using a Mitochondrial Delivery System That Inhibits the Growth of Drug-Resistant Cancer Engrafted on Mice.
Authors:Yamada Y, Munechika R, Satrialdi Kubota F, Sato Y, Sakurai Y, Harashima H
Journal:J Pharm Sci
PubMed ID:32376272
'Mitochondrial delivery of an anticancer drug targeting cancer cells would eventually result in cell death. To achieve this, a drug delivery system targeting mitochondria is needed. We recently developed a MITO-Porter, a liposome that delivers its cargo to mitochondria. We reported that such a MITO-Porter could deliver doxorubicin (DOX), an ... More
Zidovudine-Mediated Autophagy Inhibition Enhances Mitochondrial Toxicity in Muscle Cells.
Authors:Lin H, Stankov MV, Hegermann J, Budida R, Panayotova-Dimitrova D, Schmidt RE, Behrens GMN,
Journal:Antimicrob Agents Chemother
PubMed ID:30373793
Nucleoside reverse transcriptase inhibitors (NRTI), such as zidovudine (AZT), are constituents of HIV-1 therapy and are used for the prevention of mother-to-child transmission. Prolonged thymidine analogue exposure has been associated with mitochondrial toxicities to heart, liver, and skeletal muscle. We hypothesized that the thymidine analogue AZT might interfere with autophagy ... More
Lectin-induced oxidative stress in human platelets.
Authors:Signorello MG, Ravera S, Leoncini G
Journal:Redox Biol
PubMed ID:32063518
Previously we have shown that wheat germ agglutinin (WGA) and, with minor potency, Phaseolus vulgaris agglutinin (PHA), but not lens culinarian agglutinin (LCA), induce platelet aggregation, through the PLC?2 activation by the concerted action of src/syk and PI3K/BTK pathways. In this study, we have investigated platelet oxidative stress induced by ... More
Rapid Death of Follicular B Cells and Burkitt Lymphoma Cells Effectuated by Xbp1s.
Authors:Chen YT, Kung JT
Journal:J Immunol
PubMed ID:32376649
BCR-mediated tonic signaling is an indispensable requirement for the survival of follicular B (FOB) cells and Burkitt lymphoma (BL) cells. FOB cells of the I-A