Human Vascular Smooth Muscle Cell Basal Medium (formerly Medium 231)
Human Vascular Smooth Muscle Cell Basal Medium (formerly “Medium 231”) is a sterile, liquid culture medium intended for use asRead more
Have Questions?
Catalog Number
Quantity
M231500
500 mL
Catalog number M231500
Price (MXN)
-
Quantity:
500 mL
Customize this product
Human Vascular Smooth Muscle Cell Basal Medium (formerly “Medium 231”) is a sterile, liquid culture medium intended for use as one component in a complete culture environment for the growth of normal human vascular smooth muscle cells. When supplemented with SMGS, Human Vascular Smooth Muscle Cell Basal Medium will support the plating and proliferation of smooth muscle cells at densities between 2.5 x 103 cells/cm2 and 1 x 105 cells/cm2. Additional applications for use may include primary isolation of human smooth muscle cells.
Human Vascular Smooth Muscle Cell Basal Medium is a basal medium containing essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. This medium does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. This medium is HEPES and bicarbonate buffered and designed for use in an incubator with an atmosphere of 5% CO2/95% air. To support the plating and proliferation of normal human vascular smooth muscle cells, Medium 231 must be supplemented with Smooth Muscle Growth Supplement (SMGS). To induce differentiation of normal human vascular smooth muscle cells, Human Vascular Smooth Muscle Cell Basal Medium must be supplemented with Smooth Muscle Differentiation Supplement (SMDS).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeSmooth Muscle Cells
Product LineCascade Biologics
Product TypeBasal Medium
Quantity500 mL
Shipping ConditionRoom Temperature
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesHEPES
Without AdditivesNo Phenol Red
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.
Frequently asked questions (FAQs)
What is the concentration of glucose in Medium 231?
The concentration of glucose in Medium 231 is 4.6 mM.