Microplates for Fluorescence-based Assays
Invitrogen™
Microplates for Fluorescence-based Assays
Microplates for fluorescence-based assays are 96- and 384-well, black-walled, clear-bottom microplates that are optimized for fluorescence-based applications on standard fluorescence microplate readers.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoN.° de pocillosCantidad
M330909630 placas
M330899610 placas
M3309138410 placas
Número de catálogo M33090
Precio (CLP)
623.378
30 microplates
N.° de pocillos:
96
Cantidad:
30 placas
Precio (CLP)
623.378
30 microplates
Microplates for fluorescence-based assays are black-walled, clear-bottom microplates that minimize well-to-well crosstalk and autofluorescence. These 96- and 384-well microplates are compatible with standard fluorescence microplate readers and optimized for fluorescence-based applications including nucleic acid quantitation (e.g., Quant-iT assays, including PicoGreen, RiboGreen, and OliGreen).
Features of the microplates for fluorescence-based assays include:
  • Black-walled design—minimizes well-to-well crosstalk and autofluorescence
  • Clear-bottom wells—allows for optimal fluorescence detection
  • Compatibility—suitable for standard fluorescence microplate readers
  • Optimized for fluorescence microplate assays—ideal for nucleic acid quantitation (e.g., Quant-iT assays)
  • Versatile formats—available in 96- and 384-well configurations
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Esterilizables en autoclaveNot Autoclavable
DesechablesSingle-use
Para utilizar con (aplicación)Nucleic Acid Quantification
FormatoStandard
Envase30-plates
Tipo de productoMicroplates
Cantidad30 placas
Volumen de trabajo (métrico)200 μL
matriz, red, conjunto8 x 12
Código de barrasNo Barcode
ColorClear Bottom, Black
MaterialPolystyrene
N.° de pocillos96
Línea de productosInvitrogen
EsterilidadNon-sterile
Forma del pocilloFlat Bottom (F-well)
Unit Size30 microplates
Contenido y almacenamiento
Store at room temperature.