Multiplexed Proteomics™ Phosphoprotein Gel Stain Kits (with Pro-Q™ Diamond and SYPRO™ Ruby Gel Stains)
Invitrogen™

Multiplexed Proteomics™ Phosphoprotein Gel Stain Kits (with Pro-Q™ Diamond and SYPRO™ Ruby Gel Stains)

Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
M333051 Set, 1 L per Dye
M333061 Set, 200 mL per Dye
Catalog number M33305
Price (USD)
1,176.00
Each
Quantity:
1 Set, 1 L per Dye
Request bulk or custom format
Price (USD)
1,176.00
Each

Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. This reagent, when combined with our total protein stain, SYPRO Ruby Protein Gel Stain, becomes a powerful combination for quantitative proteome analysis.

Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. This reagent, when combined with our total protein stain, SYPRO Ruby Protein Gel Stain, becomes a powerful combination for quantitative proteome analysis. This convenient Pro-Q Diamond Phosphoprotein Gel Stain and SYPRO Ruby Protein Gel Stain Pack provides each stain in 1-liter amounts (Cat. No. M-33305) or in 200-mL amounts (Cat. No. M-33306).

Features include:

  • Sensitive—Pro-Q Diamond Phosphoprotein Gel Stain offers detection level down to 1–16 ng of phosphoprotein per band
  • Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. SYPRO Ruby Gel Stain is subsequently used for total protein staining and simplifies the determination of phosphorylation level
  • Linear signal—quantify with ease as Pro-Q Diamond Phosphoprotein Gel Stain and SYPRO Ruby Gel Stain exhibit excellent linearity over three orders of magnitude, ideal for quantitation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionMultiplexed Proteomics™ Phosphoprotein Gel Stain Kit #1
Detection LocationIn-Gel Detection
Detection MethodFluorescence
Quantity1 Set, 1 L per Dye
Shipping ConditionRoom Temperature
Target MoleculeProteins (Phosphoproteins)
Label or DyeSYPRO Ruby, Pro-Q Diamond
Product LinePRO-Q, SYPRO
Product TypePhosphoprotein Gel Stain Kit
Unit SizeEach
Contents & Storage
• Pro-Q Diamond Phosphoprotein Gel Stain, 1 L
• SYPRO Ruby Protein Gel Stain, 1 L.

Store at room temperature and protect from light.

Citations & References (19)

Citations & References
Abstract
Initial analysis of the phosphoproteome of Chinese hamster ovary cells using electrophoresis.
Authors:Chen Z, Southwick K, Thulin CD
Journal:J Biomol Tech
PubMed ID:15585821
'Protein phosphorylation is a common post-translational modification of enormous biological importance. Analysis of phosphorylation at the global level should shed light on the use of this modification to regulate metabolism, signal transduction, and other processes. We have begun a proteomic analysis of phosphorylation using two-dimensional gel electrophoresis. Chinese hamster ovary ... More
Comparative proteomes of the proliferating C(2)C(12) myoblasts and fully differentiated myotubes reveal the complexity of the skeletal muscle differentiation program.
Authors:Tannu NS, Rao VK, Chaudhary RM, Giorgianni F, Saeed AE, Gao Y, Raghow R
Journal:Mol Cell Proteomics
PubMed ID:15286212
'When cultured in low serum-containing growth medium, the mouse C(2)C(12) cells exit cell cycle and undergo a well-defined program of differentiation that culminates in the formation of myosin heavy chain-positive bona fide multinucleated muscle cells. To gain an understanding into this process, we compared total, membrane- and nuclear-enriched proteins, and ... More
Characterization of dynamic and steady-state protein phosphorylation using a fluorescent phosphoprotein gel stain and mass spectrometry.
Authors:Schulenberg B, Goodman TN, Aggeler R, Capaldi RA, Patton WF
Journal:Electrophoresis
PubMed ID:15300772
'Protein phosphorylation plays a vital role in the regulation of most aspects of cellular activity, being key to propagating messages within signal transduction pathways and to modulating protein function. Pro-Q Diamond phosphoprotein gel stain is suitable for the fluorescence detection of phosphoserine-, phosphothreonine-, and phosphotyrosine-containing proteins directly in sodium dodecyl ... More
Chronophin, a novel HAD-type serine protein phosphatase, regulates cofilin-dependent actin dynamics.
Authors:Gohla A, Birkenfeld J, Bokoch GM
Journal:Nat Cell Biol
PubMed ID:15580268
Cofilin is a key regulator of actin cytoskeletal dynamics whose activity is controlled by phosphorylation of a single serine residue. We report the biochemical isolation of chronophin (CIN), a unique cofilin-activating phosphatase of the haloacid dehalogenase (HAD) superfamily. CIN directly dephosphorylates cofilin with high specificity and colocalizes with cofilin in ... More
Determination of in vivo protein phosphorylation in photosynthetic membranes.
Authors:Vainonen JP, Vener AV, Aro EM,
Journal:Methods Mol Biol
PubMed ID:19083170
Light- and redox-controlled reversible phosphorylation of thylakoid proteins regulates short- and long-term acclimation of plants to environmental cues. The major phosphoproteins in thylakoids belong to photosystem II and its light-harvesting antenna but phosphorylation of subunits of other thylakoid protein complexes has been detected as well. The detection methods include electrophoretic ... More