Thermo Scientific™
Richard-Allan Scientific™ Resolve™ Immersion Oil
 Related applications:
Anatomical Pathology
Eliminate toxicity and disposal problems with the Thermo Scientific™ Richard-Allan Scientific™ Resolve™ Immersion Oil, which contains no PCBs. It is a clear, colorless oil that allows light transmission throughout most of the UV-VIS spectrum.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
  • Packaged conveniently in four sizes
Product Details:
  • Available in both low and high viscosity
  • Non-drying and non-hardening
  • Refractive index of 1.5150
Documents
Manuals & protocols