Our facility does not allow flow cytometry of live cells. Can I fix the cells after staining with MitoTracker Red CMXRos dye?
Yes, this is possible because MitoTracker Red CMXRox dye is retained upon fixation. See Table 2 in this manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/mp07510.pdf).