L(+)-Leucine USP, Multi-Compendial, GMP, Macron Fine Chemicals™
Avantor™

L(+)-Leucine USP, Multi-Compendial, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets U.S.P, B.P., E.P and J.P Specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
M4944-061 kg
M4944-0712 kg
Número de catálogo M4944-06
Precio (MXN)
-
Cantidad:
1 kg
This product is GMP Manufactured and meets U.S.P, B.P., E.P and J.P Specifications.
This product is GMP Manufactured and meets U.S.P, B.P., E.P and J.P Specifications.
Especificaciones
CAS61-90-5
Nombre del producto químico o materialL(+)-Leucine
ColorWhite
Punto de fusión300°C
Fórmula molecularC6H13NO2
SinónimoL-Leucine, (S)-(+)-Leucine, Leucine, Leu, H-Leu-OH
GradoMulti-Compendial
Peso molecular (g/mol)131.17
Porcentaje de pureza98.5-101.5%
Cantidad1 kg
Unit SizeEach