Malachite Green isothiocyanate
Invitrogen™

Malachite Green isothiocyanate

malachite green isothiocyanate
Have Questions?
Catalog NumberQuantity
M68910 mg
Catalog number M689
Price (USD)
502.00
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
502.00
Each

Malachite green is a nonfluorescent photosensitizer that absorbs at long wavelengths (∼630 nm). Its photosensitizing action can be targeted to particular cellular sites by using the amine-reactive malachite green isothiocyanate to conjugate this label to antibodies or other biomolecules.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Chemical ReactivityAmine
Label or DyeMalchite Green
Product TypeMalachite Green Isothiocyanate
Quantity10 mg
Reactive MoietyIsothiocyanate
Shipping ConditionRoom Temperature
ColorGreen
Label TypeOther Labels or Dyes
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (14)

Citations & References
Abstract
Talin and vinculin play distinct roles in filopodial motility in the neuronal growth cone.
Authors:Sydor AM, Su AL, Wang FS, Xu A, Jay DG
Journal:J Cell Biol
PubMed ID:8794861
'Filopodial motility is critical for many biological processes, particularly for axon guidance. This motility is based on altering the F-actin-based cytoskeleton, but the mechanisms of how this occurs and the actin-associated proteins that function in this process remain unclear. We investigated two of these proteins found in filopodia, talin and ... More
Chromophore-assisted laser inactivation of a repulsive axonal guidance molecule.
Authors:Müller BK, Jay DG, Bonhoeffer F
Journal:Curr Biol
PubMed ID:8939610
'BACKGROUND: The axons of retinal ganglion neurons from a precise topographic map in the optic tectum in the midbrain, and the guidance of retinal axons by directional cues in the tectum is crucial in this process. Several in vitro systems have been developed in order to identify the molecular basis ... More
Photosensitization of Staphylococcus aureus with malachite green isothiocyanate: inactivation efficiency and spectroscopic analysis.
Authors:Golding PS, King TA, Maddocks L, Drucker DB, Blinkhorn AS
Journal:J Photochem Photobiol B
PubMed ID:10093920
'The potential of malachite green isothiocyanate as a photosensitizer for the inactivation of bacteria has been evaluated. Samples of Staphylococcus aureus are treated with the dye and exposed to continuous-wave red light from a filtered xenon lamp. Reduction in cell viability is seen to increase with radiation dose, whilst non-photosensitized ... More
Spatial specificity of chromophore assisted laser inactivation of protein function.
Authors:Linden KG, Liao JC, Jay DG
Journal:Biophys J
PubMed ID:1581504
'Chromophore assisted laser inactivation (CALI) is a new technique that selectively inactivates proteins of interest to elucidate their in vivo functions. This method has application to a wide array of biological questions. An understanding of aspects of the mechanism of CALI is required for its judicious application. A critical concern ... More
Chromophore-assisted laser inactivation of cellular proteins.
Authors:Beermann AE, Jay DG
Journal:Methods Cell Biol
PubMed ID:7707977