Trisodium citrate, dihydrate, 99.0-100.5% (by anhydrous basis), crystals, GenAR™, USP, Multi-Compendial, GMP, Macron Fine Chemicals™
Avantor™

Trisodium citrate, dihydrate, 99.0-100.5% (by anhydrous basis), crystals, GenAR™, USP, Multi-Compendial, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Catalog number M7773-20
Price (MXN)
-
Quantity:
100 lb.
This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Specifications
CAS4-3-6132
Chemical Name or MaterialTrisodium citrate dihydrate
ColorWhite
Melting Point150°C
Molecular FormulaHOC(COONa)(CH2COONa)2·2H2O
SynonymCitric acid tridosium salt dihydrate, Sodium citrate dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate
GradeMulti-Compendial
Molecular Weight (g/mol)294.1
Percent Purity99.0-100.5%
Quantity100 lb.
Unit Size100 lb.