D-(-)-Mannitol, 97.0-102.0%, powder, GenAR™, USP for biotechnology, GMP, Macron Fine Chemicals™
Avantor™

D-(-)-Mannitol, 97.0-102.0%, powder, GenAR™, USP for biotechnology, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets USP, BP, and EP Requirements.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
M7781-04500 g
7781-8812 kg
Número de catálogo M7781-04
Precio (CLP)
-
Cantidad:
500 g
This product is GMP Manufactured and meets USP, BP, and EP Requirements.
Especificaciones
Punto de ebullición290°C to 295°C (4 hPa)
GradoBiotech
Peso molecular (g/mol)182.17
Porcentaje de pureza97.0-102.0%
Forma físicaPowder
Cantidad500 g
Unit Size500 g