Thermo Scientific™
Vari-Mix™ Platform Rocker
 Related applications:
Lab Shakers
Utilize the steep angle rocking of the Thermo Scientific™ Vari-Mix™ Platform Rocker for applications such as hybridization, blotting and staining or destaining gels. Variable speed control provides a gentle to vigorous wave motion. The built-in timer makes it easy to perform time-dependent studies.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Features:
  • Large white nonskid rubber mixing platform easily adjusts angle of motion
  • Timer can be set for continuous or timed operation
  • Rocker can be used in many different laboratory applications
  • Platform removes easily for autoclaving and allows easy viewing of contents
  • All finishes are chemical-resistant and easy to clean
  • Optional double-tier platform doubles mixing surface
Includes:
3-wire cord and plug; 240V model also includes European cord set
Compatible with:
  • Double-tier platform (AY797X1)
  • Adjustable Labware Platform (AY491X1A)
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Documents
Manuals & protocols