WebSeal™ Microplate Sealing System
Actual product may vary
Thermo Scientific™
WebSeal™ Microplate Sealing System
The Thermo Scientific™ National WebSeal Microplate Sealing System provides superior resealability after multiple injections.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsWell ShapeColor
M9601-RD1DB96RoundDark Blue
M3841-R1R384RoundRed
Catalog number M9601-RD1DB
Price (USD)
-
No. of Wells:
96
Well Shape:
Round
Color:
Dark Blue
The Thermo Scientific™ National WebSeal Microplate Sealing System provides superior resealability after multiple injections.
When productivity really counts and cross-contamination simply won't be tolerated, choose the WebSeal System for efficiency and security. WebSeal system products are used in high-throughput screening, combinatorial chemistry, life science and HPLC. This system is a comprehensive range of 96-well microtiter plates with glass inserts and silicone/PTFE sealing mats.
  • Manufactured from pure silicone with PTFE coating on underside
  • Excellent chemical compatibility
  • Dry heat autoclavable
  • Eliminates cross-contamination well to well
  • Available pre-slit for easy penetration and reduced vacuum formation
Specifications
No. per Pack5
Product TypeMicroplate Sealing Components
ColorDark Blue
MaterialPTFE-coated Silicone
No. of Wells96
Plate Compatibility96-well Plate
Product LineWebseal
Well ShapeRound
Unit SizeEach